Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmya5
Entrez Gene ID
688915
Gene Name
cardiomyopathy associated 5
Gene Aliases
-
Chromosome Location
Chr.2: 22646694 - 22744533 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006231789.2 XP_006231851.1 11 - 12 11625 114

View Full Product Details