The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039022.1 | NP_001034111.1 | 2 - 3 | 352 | 107 |
GenBank mRNA | BC098719.1 | - | 2 - 3 | 352 | 107 |
FQ219423.1 | - | 2 - 3 | 357 | 107 |