Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chchd3
Entrez Gene ID
296966
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 60080538 - 60358562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106588.1 NP_001100058.1 2 - 3 328 111
GenBank mRNA FQ216356.1 - 2 - 3 311 111

View Full Product Details