Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcrb
Entrez Gene ID
24820
Gene Name
T-cell receptor beta chain
Gene Aliases
RATTCB, RATTCBC1, TCB, TCBC1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC091277.1 - - 835 87
BC091428.1 - - 921 87
BC099166.1 - - 963 87
BC105831.1 - - 833 87
BC107948.1 - - 778 87
BC158832.1 - - 831 87
X14319.1 - - 657 87

View Full Product Details