Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptar1
Entrez Gene ID
286972
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 241501679 - 241557651 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105760.2 NP_001099230.2 1 - 2 96 78

View Full Product Details