Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arc
Entrez Gene ID
54323
Gene Name
activity-regulated cytoskeleton-associated protein
Gene Aliases
rg3.1
Chromosome Location
Chr.7: 115907097 - 115911054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA Z46925.1 - 2 - 3 64

View Full Product Details