The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127295.2 | NP_001120767.1 | 2 - 3 | 155 | 108 |
GenBank mRNA | BC167088.1 | - | 2 - 3 | 155 | 108 |
FQ220736.1 | - | 2 - 3 | 189 | 108 | |
FQ225628.1 | - | 2 - 3 | 155 | 108 | |
FQ229831.1 | - | 2 - 3 | 190 | 108 |