Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ext1
Entrez Gene ID
299907
Gene Name
exostosin glycosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.7: 92605008 - 92881392 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130540.1 NP_001124012.1 2 - 3 1180 72
GenBank mRNA BC105839.1 - 2 - 3 495 72

View Full Product Details