Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl6
Entrez Gene ID
362941
Gene Name
F-box and leucine-rich repeat protein 6
Gene Aliases
-
Chromosome Location
Chr.7: 117602056 - 117604954 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005563.1 NP_001005563.1 4 - 5 758 68
GenBank mRNA BC082108.1 - 4 - 5 758 68

View Full Product Details