Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eef1d
Entrez Gene ID
300033
Gene Name
eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein)
Gene Aliases
-
Chromosome Location
Chr.7: 116928264 - 116942981 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765555.1 XP_008763777.1 2 - 3 1348 62
XM_008765560.1 XP_008763782.1 2 - 3 224 62

View Full Product Details