Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il27
Entrez Gene ID
365368
Gene Name
interleukin 27
Gene Aliases
RGD1561420
Chromosome Location
Chr.1: 198003622 - 198010112 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_344962.7 XP_344963.5 1 - 2 1249 61

View Full Product Details