The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024360.2 | NP_001019531.1 | 5 - 6 | 984 | 68 |
XM_008766878.1 | XP_008765100.1 | 4 - 5 | 1960 | 68 | |
GenBank mRNA | AB210855.1 | - | 5 - 6 | 884 | 68 |
BC128712.1 | - | 5 - 6 | 984 | 68 |