Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsph9
Entrez Gene ID
316238
Gene Name
radial spoke head 9 homolog (Chlamydomonas)
Gene Aliases
RGD1310693
Chromosome Location
Chr.9: 17224600 - 17245093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108205.1 NP_001101675.1 4 - 5 722 88

View Full Product Details