The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013131.1 | NP_037263.1 | 5 - 6 | 2834 | 75 |
GenBank mRNA | M36074.1 | - | 5 - 6 | 2834 | 75 |