The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133310.2 | NP_579844.2 | 4 - 5 | 728 | 63 |
GenBank mRNA | AF075595.1 | - | 4 - 5 | 516 | 63 |
BC083923.1 | - | 5 - 6 | 933 | 63 |