The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_078620.1 | NP_511175.1 | 7 - 8 | 3241 | 78 |
GenBank mRNA | U53420.1 | - | 7 - 8 | 3241 | 78 |