Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sytl1
Entrez Gene ID
297872
Gene Name
synaptotagmin-like 1
Gene Aliases
Slp-1
Chromosome Location
Chr.5: 151448639 - 151459875 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025651.1 NP_001020822.1 8 - 9 833 122
XM_006239013.2 XP_006239075.1 8 - 9 1671 122
GenBank mRNA BC098679.1 - 8 - 9 833 122

View Full Product Details