Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mad2l2
Entrez Gene ID
313702
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Gene Aliases
-
Chromosome Location
Chr.5: 164943203 - 164956255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012106.1 NP_001012106.1 1 - 2 112 64
XM_006239390.2 XP_006239452.1 3 - 4 281 64
XM_006239391.2 XP_006239453.1 1 - 2 797 64
XM_008764256.1 XP_008762478.1 2 - 3 518 64
GenBank mRNA BC087687.1 - 1 - 2 112 64

View Full Product Details