The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079698.1 | NP_001073166.1 | 5 - 6 | 706 | 58 |
XM_006254917.2 | XP_006254979.1 | 5 - 6 | 995 | 58 | |
GenBank mRNA | BC087117.1 | - | 4 - 5 | 946 | 58 |
BC128781.1 | - | 5 - 6 | 706 | 58 |