The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035238.2 | NP_001030315.1 | 6 - 7 | 634 | 66 |
NM_031021.3 | NP_112283.1 | 6 - 7 | 685 | 66 | |
XM_006256081.2 | XP_006256143.1 | 6 - 7 | 721 | 66 | |
GenBank mRNA | BC078807.1 | - | 6 - 7 | 634 | 66 |
EV778558.1 | - | 6 - 7 | 689 | 66 | |
FQ230013.1 | - | 6 - 7 | 673 | 66 | |
HE864426.1 | - | 5 - 6 | 569 | 66 | |
HE864432.1 | - | - | 569 | 66 | |
HE864433.1 | - | - | 569 | 66 | |
HE864434.1 | - | - | 569 | 66 |