Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pbx2
Entrez Gene ID
406164
Gene Name
pre-B-cell leukemia homeobox 2
Gene Aliases
-
Chromosome Location
Chr.20: 4357805 - 4362730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002828.2 NP_001002828.1 3 - 4 624 81

View Full Product Details