Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd4
Entrez Gene ID
293856
Gene Name
PDZ domain containing 4
Gene Aliases
RGD1561214
Chromosome Location
Chr.X: 156963329 - 156993591 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135836.1 NP_001129308.1 3 - 4 385 154
XM_006229562.2 XP_006229624.1 3 - 4 944 154
XM_006229563.2 XP_006229625.1 3 - 4 795 154
XM_006229564.2 XP_006229626.1 3 - 4 944 154

View Full Product Details