Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atf1
Entrez Gene ID
315305
Gene Name
activating transcription factor 1
Gene Aliases
Atf-1
Chromosome Location
Chr.7: 141882261 - 141924790 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100895.1 NP_001094365.1 3 - 4 457 118
GenBank mRNA BC091222.1 - 3 - 4 431 118
BC105853.1 - 3 - 4 454 118
BC168224.1 - 3 - 4 458 118
CO565801.1 - - 457 118

View Full Product Details