Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtsf1
Entrez Gene ID
315347
Gene Name
gametocyte specific factor 1
Gene Aliases
RGD1310791
Chromosome Location
Chr.7: 145030139 - 145049180 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765751.1 XP_008763973.1 1 - 2 81 94

View Full Product Details