Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtsf1
Entrez Gene ID
315347
Gene Name
gametocyte specific factor 1
Gene Aliases
RGD1310791
Chromosome Location
Chr.7: 145030139 - 145049180 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079707.1 NP_001073175.1 5 - 6 536 103
XM_006242403.2 XP_006242465.1 6 - 7 508 103
XM_008765750.1 XP_008763972.1 6 - 7 498 103
XM_008765751.1 XP_008763973.1 6 - 7 500 103
XM_008765752.1 XP_008763974.1 6 - 7 569 103
GenBank mRNA BC128778.1 - 5 - 6 536 103

View Full Product Details