Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2r2a
Entrez Gene ID
117104
Gene Name
protein phosphatase 2, regulatory subunit B, alpha
Gene Aliases
-
Chromosome Location
Chr.15: 43673958 - 43733182 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053999.2 NP_446451.1 3 - 4 489 103
XM_008770730.1 XP_008768952.1 3 - 4 1296 103
XM_008770731.1 XP_008768953.1 3 - 4 1296 103
GenBank mRNA BC128731.1 - 3 - 4 489 103
M83298.1 - 3 - 4 470 103

View Full Product Details