Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnma1
Entrez Gene ID
83731
Gene Name
potassium channel, calcium activated large conductance subfamily M alpha, member 1
Gene Aliases
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
Chromosome Location
Chr.15: 344204 - 1048849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031828.1 NP_114016.1 8 - 9 1627 157
GenBank mRNA AB248960.1 - 1 - 2 43 157
AF083341.1 - 8 - 9 1063 157
AF135265.1 - 8 - 9 1481 157
AY330290.1 - 8 - 9 952 157
AY330291.1 - 8 - 9 952 157
AY330292.1 - 8 - 9 961 157
AY330293.2 - 8 - 9 952 157
AY330294.1 - 8 - 9 952 157
AY344964.1 - 8 - 9 952 157
U40603.1 - 8 - 9 1627 157
U55995.1 - 8 - 9 1105 157
U93052.1 - 8 - 9 1053 157

View Full Product Details