Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vnn3
Entrez Gene ID
498992
Gene Name
vanin 3
Gene Aliases
RGD1560609
Chromosome Location
Chr.1: 22649114 - 22661376 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109136.1 NP_001102606.1 6 - 7 1498 107

View Full Product Details