The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012966.1 | NP_037098.1 | - | 526 | 159 |
GenBank mRNA | BC058492.1 | - | - | 301 | 159 |
FQ209877.1 | - | - | 314 | 159 | |
FQ220992.1 | - | - | 338 | 159 | |
FQ222153.1 | - | - | 313 | 159 | |
X71429.1 | - | - | 272 | 159 |