Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp2b1
Entrez Gene ID
29598
Gene Name
ATPase, Ca++ transporting, plasma membrane 1
Gene Aliases
Pmca1a, Pmca1b, Pmca1c
Chromosome Location
Chr.7: 41114606 - 41223116 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053311.1 NP_445763.1 9 - 10 1609 65
XM_006241294.2 XP_006241356.1 9 - 10 1701 65
XM_006241295.2 XP_006241357.1 8 - 9 1578 65
XM_006241296.2 XP_006241358.1 9 - 10 1701 65
GenBank mRNA J03753.1 - 9 - 10 1609 65

View Full Product Details