The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109204.1 | NP_001102674.1 | 9 - 10 | 1109 | 65 |
XM_006234784.2 | XP_006234846.1 | 10 - 11 | 1287 | 65 | |
GenBank mRNA | BC168875.1 | - | 9 - 10 | 1073 | 65 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008762135.1 | XP_008760357.1 | 5 - 6 | 646 | 65 |