Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptlc3
Entrez Gene ID
296188
Gene Name
serine palmitoyltransferase, long chain base subunit 3
Gene Aliases
RGD1310030
Chromosome Location
Chr.3: 132560437 - 132689313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106517.1 NP_001099987.1 1 - 2 403 100

View Full Product Details