Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fastkd1
Entrez Gene ID
311112
Gene Name
FAST kinase domains 1
Gene Aliases
RGD1563531
Chromosome Location
Chr.3: 55925598 - 55951584 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191738.1 NP_001178667.1 10 - 11 3042 76
XM_006234311.1 XP_006234373.1 10 - 11 1851 76

View Full Product Details