Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Larp4b
Entrez Gene ID
307070
Gene Name
La ribonucleoprotein domain family, member 4B
Gene Aliases
Larp5, RGD1310713
Chromosome Location
Chr.17: 63236007 - 63317298 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107361.1 NP_001100831.1 11 - 12 1219 91

View Full Product Details