Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcea3
Entrez Gene ID
298559
Gene Name
transcription elongation factor A (SII), 3
Gene Aliases
-
Chromosome Location
Chr.5: 154598765 - 154634687 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015008.1 NP_001015008.1 3 - 4 326 145
GenBank mRNA BC091180.1 - 3 - 4 326 145

View Full Product Details