Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gsdma
Entrez Gene ID
360619
Gene Name
gasdermin A
Gene Aliases
Gsdma1
Chromosome Location
Chr.10: 86569903 - 86595102 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108297.1 NP_001101767.1 2 - 3 308 52
XM_006247347.1 XP_006247409.1 2 - 3 343 52
GenBank mRNA BC112385.1 - 2 - 3 292 52

View Full Product Details