Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vezf1
Entrez Gene ID
287615
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.10: 75365811 - 75382012 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768166.1 XP_008766388.1 5 - 6 1248 69
XM_008768167.1 XP_008766389.1 5 - 6 1230 69
GenBank mRNA FQ108271.1 - 4 - 5 550 69

View Full Product Details