Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbx2
Entrez Gene ID
303730
Gene Name
chromobox homolog 2
Gene Aliases
-
Chromosome Location
Chr.10: 108132105 - 108140935 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107071.1 NP_001100541.1 4 - 5 339 74

View Full Product Details