The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035007.1 | NP_001030179.1 | 10 - 11 | 1429 | 87 |
XM_006256944.2 | XP_006257006.1 | 8 - 9 | 1118 | 87 | |
GenBank mRNA | BC097391.1 | - | - | 1391 | 87 |