Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a37
Entrez Gene ID
306000
Gene Name
solute carrier family 25 (mitochondrial iron transporter), member 37
Gene Aliases
Mscp, RGD1359361
Chromosome Location
Chr.15: 51128027 - 51168493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013996.1 NP_001014018.1 3 - 4 640 66
XM_006252266.1 XP_006252328.1 4 - 5 812 66
GenBank mRNA BC082071.1 - 3 - 4 640 66
FQ227341.1 - 3 - 4 581 66
FQ230701.1 - 2 - 3 297 66
FQ231671.1 - 2 - 3 111 66
FQ231945.1 - 2 - 3 128 66
FQ233331.1 - 3 - 4 540 66

View Full Product Details