Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcidas
Entrez Gene ID
688802
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Gene Aliases
Idas, Mci, Mcin, Multicilin
Chromosome Location
Chr.2: 44863246 - 44869692 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001068366.4 XP_001068366.2 6 - 7 766 85

View Full Product Details