The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034011.1 | NP_001029183.1 | 3 - 4 | 240 | 60 |
GenBank mRNA | BC099092.1 | - | 3 - 4 | 240 | 60 |