The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107572.1 | NP_001101042.1 | 2 - 3 | 630 | 81 |
XM_006230794.2 | XP_006230856.1 | 2 - 3 | 2654 | 81 | |
XM_006230796.2 | XP_006230858.1 | 2 - 3 | 1314 | 81 | |
XM_008760138.1 | XP_008758360.1 | 1 - 2 | 1770 | 81 |