Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmaa
Entrez Gene ID
291939
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
-
Chromosome Location
Chr.19: 32294087 - 32325927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106174.1 NP_001099644.1 3 - 4 854 107
XM_006255398.2 XP_006255460.1 2 - 3 536 107
XM_006255399.2 XP_006255461.1 2 - 3 2675 107

View Full Product Details