The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191759.1 | NP_001178688.1 | 6 - 7 | 857 | 65 |
XM_006238495.2 | XP_006238557.1 | 7 - 8 | 1177 | 65 | |
GenBank mRNA | BC090023.1 | - | 3 - 4 | 382 | 65 |