Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stoml2
Entrez Gene ID
298203
Gene Name
stomatin (Epb7.2)-like 2
Gene Aliases
-
Chromosome Location
Chr.5: 58472561 - 58476158 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031646.1 NP_001026816.1 3 - 4 275 84
GenBank mRNA BC099164.1 - 3 - 4 275 84
FQ213146.1 - 3 - 4 306 84

View Full Product Details