The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013227.2 | NP_001013245.1 | 7 - 8 | 603 | 84 |
GenBank mRNA | BC063182.1 | - | 7 - 8 | 607 | 84 |
FQ215552.1 | - | 7 - 8 | 602 | 84 |