Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankdd1a
Entrez Gene ID
100362785
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
Chromosome Location
Chr.8: 71034630 - 71057264 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243300.2 XP_006243362.1 11 - 12 2337 78

View Full Product Details