Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Socs5
Entrez Gene ID
500616
Gene Name
suppressor of cytokine signaling 5
Gene Aliases
RGD1564914
Chromosome Location
Chr.6: 10671323 - 10705610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109274.1 NP_001102744.1 1 - 2 358 73

View Full Product Details