Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc34
Entrez Gene ID
366517
Gene Name
tetratricopeptide repeat domain 34
Gene Aliases
RGD1304840
Chromosome Location
Chr.5: 172253257 - 172269310 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_345615.6 XP_345616.5 5 - 6 2206 67

View Full Product Details